missing translation for 'onlineSavingsMsg'
Learn More

O6-Benzylguanine, 98%

CAS: 19916-73-5 | C12H11N5O | 241.25 g/mol

Supplier:  Thermo Scientific Chemicals H60274MD

Catalog No. AAH60274MD


Only null left
Explore available promotions

Description

Description

O6-Benzylguanine is an effective substrate and efficient inactivator of DNA alkyltransferase. O6-Benzylguanine is an inhibitor of MGMT. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O6-Benzylguanine is an effective substrate and efficient inactivator of DNA alkyltransferase. O6-Benzylguanine is an inhibitor of MGMT. It is also used as an important raw material and intermediate used in organic Synthesis, pharmac
Specifications

Specifications

O6-Benzylguanine
C12H11N5O
250 mg
Soluble in methanol (20mg/ml),and aqueous alkali (Heated).
NC1=NC(OCC2=CC=CC=C2)=C2NC=NC2=N1
241.25
241.25
19916-73-5
MFCD00269931
o6-benzylguanine, 6-o-benzylguanine, 6-benzyloxy-7h-purin-2-amine, o 6-benzylguanine, 2-amino-6-benzyloxypurine, 6-benzyloxy-9h-purin-2-amine, o 6-bgua, 6-benzyloxy guanine, 2-amino-6-benzyloxy purine, 6-benzyloxyguanine
KRWMERLEINMZFT-UHFFFAOYSA-N
6-phenylmethoxy-7H-purin-2-amine
4578
98%

Chemical Identifiers

19916-73-5
241.25
KRWMERLEINMZFT-UHFFFAOYSA-N
4578
NC1=NC(OCC2=CC=CC=C2)=C2NC=NC2=N1
C12H11N5O
MFCD00269931
o6-benzylguanine, 6-o-benzylguanine, 6-benzyloxy-7h-purin-2-amine, o 6-benzylguanine, 2-amino-6-benzyloxypurine, 6-benzyloxy-9h-purin-2-amine, o 6-bgua, 6-benzyloxy guanine, 2-amino-6-benzyloxy purine, 6-benzyloxyguanine
6-phenylmethoxy-7H-purin-2-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only