CAS RN 19916-73-5
IUPAC Name:
6-(benzyloxy)-7H-purin-2-amine
Synonyms:
6-O-benzylguanine
Molecular Weight (g/mol):
241.25
Molecular Formula:
C12H11N5O
InChi Key:
KRWMERLEINMZFT-UHFFFAOYSA-N
SMILES:
NC1=NC(OCC2=CC=CC=C2)=C2NC=NC2=N1
1
–
1
of
1
results
1
Promotions Available
O6-Benzylguanine, 98%
O6-Benzylguanine, 98%
CAS: 19916-73-5 Molecular Formula: C12H11N5O Molecular Weight (g/mol): 241.25 MDL Number: MFCD00269931 InChI Key: KRWMERLEINMZFT-UHFFFAOYSA-N Synonym: o6-benzylguanine,6-o-benzylguanine,6-benzyloxy-7h-purin-2-amine,o 6-benzylguanine,2-amino-6-benzyloxypurine,6-benzyloxy-9h-purin-2-amine,o 6-bgua,6-benzyloxy guanine,2-amino-6-benzyloxy purine,6-benzyloxyguanine PubChem CID: 4578 IUPAC Name: 6-phenylmethoxy-7H-purin-2-amine SMILES: NC1=NC(OCC2=CC=CC=C2)=C2NC=NC2=N1