missing translation for 'onlineSavingsMsg'
Learn More

o-Xylylene dichloride, 98%

Catalog No. AAA1250030
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1250030 250 g
AAA1250018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1250030 Supplier Thermo Scientific Chemicals Supplier No. A1250030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 612-12-4 | C8H8Cl2 | 175.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 612-12-4
Molecular Formula C8H8Cl2
Molecular Weight (g/mol) 175.05
MDL Number MFCD00000903
InChI Key FMGGHNGKHRCJLL-UHFFFAOYSA-N
Synonym 1,2-bis chloromethyl benzene, o-xylylene dichloride, alpha,alpha'-dichloro-o-xylene, o-bis chloromethyl benzene, benzene, 1,2-bis chloromethyl, ccris 1773, o-xylene, alpha,alpha'-dichloro, benzene, 1,2-dichloromethyl, .alpha.,.alpha.'-dichloro-o-xylene, o-xylylenchlorid
PubChem CID 11919
IUPAC Name 1,2-bis(chloromethyl)benzene
SMILES ClCC1=CC=CC=C1CCl

Specifications

Melting Point 53°C to 57°C
Boiling Point 246°C
Flash Point 107°C (224°F)
Odor Pungent
Quantity 250 g
UN Number UN2928
Beilstein 2043675
Formula Weight 175.06
Percent Purity 98%
Chemical Name or Material o-Xylylene dichloride
Hazard Category H302-H314-H330
Hazard Statement GHS H Statement
H330-H314-H318-H302
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 210-291-8
RTECSNumber ZE4085000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only