missing translation for 'onlineSavingsMsg'
Learn More

o-Xylyl Cyanide 98.0+%, TCI America™

Catalog No. X004525ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X004525ML 25 mL
X0045100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X004525ML Supplier TCI America Supplier No. X004525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22364-68-7
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00001904
InChI Key WMGVPDQNPUQRND-UHFFFAOYSA-N
Synonym 2-methylbenzyl cyanide, 2-o-tolyl acetonitrile, o-tolylacetonitrile, 2-methylphenylacetonitrile, o-methylphenylacetonitrile, o-xylyl cyanide, o-methylbenzyl cyanide, acetonitrile, o-tolyl, benzeneacetonitrile, 2-methyl, 2-2-methylphenyl acetonitrile
PubChem CID 31155
ChEBI CHEBI:27982
IUPAC Name 2-(2-methylphenyl)acetonitrile
SMILES CC1=CC=CC=C1CC#N

Specifications

Color Yellow
Boiling Point 120°C
Quantity 25 mL
UN Number 3276
Formula Weight 131.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material o-Xylyl Cyanide
RTECSNumber AM2290000
TSCA No