missing translation for 'onlineSavingsMsg'
Learn More

o-Tolylthiourea 98.0+%, TCI America™

Catalog No. T065725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T065725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T065725G Supplier TCI America Supplier No. T065725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 614-78-8
Molecular Formula C8H10N2S
Molecular Weight (g/mol) 166.24
MDL Number MFCD00041162
InChI Key ACLZYRNSDLQOIA-UHFFFAOYSA-N
Synonym o-tolylthiourea, 2-methylphenyl thiourea, n-o-tolyl thiourea, 1-2-methylphenyl thiourea, n-2-methylphenyl thiourea, 1-2-methylphenyl-2-thiourea, 2-tolylthiourea, 1-o-tolyl-2-thiourea, thiourea, 2-methylphenyl, 1-o-tolyl thiourea
PubChem CID 2787703
IUPAC Name (2-methylphenyl)thiourea
SMILES CC1=CC=CC=C1NC(N)=S

Specifications

Melting Point 156°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 166.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material o-Tolylthiourea
EINECSNumber (3)-2207
RTECSNumber YU2975000
TSCA Yes