Learn More
o-Tolylacetonitrile, 98+%
CAS: 22364-68-7 | C9H9N | 131.178 g/mol
Supplier: Thermo Scientific Chemicals A1478714
Description
o-Tolylacetonitrile is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic and chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationso-Tolylacetonitrile is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic and chemical intermediate.
Solubility
Slightly soluble in water. Soluble in benzene, ethyl ether, ethanol.
Notes
Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
Specifications
| o-Tolylacetonitrile | |
| 1.01 | |
| >110°C (230°F) | |
| C9H9N | |
| MFCD00001904 | |
| UN3276 | |
| 2-methylbenzyl cyanide, 2-o-tolyl acetonitrile, o-tolylacetonitrile, 2-methylphenylacetonitrile, o-methylphenylacetonitrile, o-xylyl cyanide, o-methylbenzyl cyanide, acetonitrile, o-tolyl, benzeneacetonitrile, 2-methyl, 2-2-methylphenyl acetonitrile | |
| WMGVPDQNPUQRND-UHFFFAOYSA-N | |
| 2-(2-methylphenyl)acetonitrile | |
| 31155 | |
| 131.18 |
| 22364-68-7 | |
| 211°C to 213°C | |
| Almond-like | |
| 1.5275 | |
| 25 g | |
| 907182 | |
| Slightly soluble in water. Soluble in benzene,ethyl ether,ethanol. | |
| CC1=CC=CC=C1CC#N | |
| 131.178 | |
| CHEBI:27982 | |
| ≥98% |
Chemical Identifiers
| 22364-68-7 | |
| 131.178 | |
| WMGVPDQNPUQRND-UHFFFAOYSA-N | |
| 31155 | |
| 2-(2-methylphenyl)acetonitrile |
| C9H9N | |
| MFCD00001904 | |
| 2-methylbenzyl cyanide, 2-o-tolyl acetonitrile, o-tolylacetonitrile, 2-methylphenylacetonitrile, o-methylphenylacetonitrile, o-xylyl cyanide, o-methylbenzyl cyanide, acetonitrile, o-tolyl, benzeneacetonitrile, 2-methyl, 2-2-methylphenyl acetonitrile | |
| CHEBI:27982 | |
| CC1=CC=CC=C1CC#N |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber : 244-937-5
RTECSNumber : AM2290000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only