missing translation for 'onlineSavingsMsg'
Learn More

o-Tolylacetonitrile, 98+%

Catalog No. AAA1478706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1478706 5 g
AAA1478714 25 g
AAA1478722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1478706 Supplier Thermo Scientific Chemicals Supplier No. A1478706

CAS: 22364-68-7 | C9H9N | 131.178 g/mol

o-Tolylacetonitrile is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Tolylacetonitrile is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an organic and chemical intermediate.

Solubility
Slightly soluble in water. Soluble in benzene, ethyl ether, ethanol.

Notes
Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.

Chemical Identifiers

CAS 22364-68-7
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00001904
InChI Key WMGVPDQNPUQRND-UHFFFAOYSA-N
Synonym 2-methylbenzyl cyanide, 2-o-tolyl acetonitrile, o-tolylacetonitrile, 2-methylphenylacetonitrile, o-methylphenylacetonitrile, o-xylyl cyanide, o-methylbenzyl cyanide, acetonitrile, o-tolyl, benzeneacetonitrile, 2-methyl, 2-2-methylphenyl acetonitrile
PubChem CID 31155
ChEBI CHEBI:27982
IUPAC Name 2-(2-methylphenyl)acetonitrile
SMILES CC1=CC=CC=C1CC#N

Specifications

Density 1.01
Boiling Point 211°C to 213°C
Flash Point >110°C (230°F)
Odor Almond-like
Refractive Index 1.5275
Quantity 5 g
UN Number UN3276
Beilstein 907182
Solubility Information Slightly soluble in water. Soluble in benzene,ethyl ether,ethanol.
Formula Weight 131.18
Percent Purity ≥98%
Chemical Name or Material o-Tolylacetonitrile
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 244-937-5
RTECSNumber AM2290000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only