missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Tolyl Isothiocyanate 99.0+%, TCI America™
$121.16 - $480.92
Chemical Identifiers
| CAS | 614-69-7 |
|---|---|
| Molecular Formula | C8H7NS |
| Molecular Weight (g/mol) | 149.211 |
| MDL Number | MFCD00004802 |
| InChI Key | JYKYYPPZLPVIBY-UHFFFAOYSA-N |
| Synonym | 2-methylphenyl isothiocyanate, o-tolyl isothiocyanate, 2-tolyl isothiocyanate, o-tolylisothiocyanate, benzene, 1-isothiocyanato-2-methyl, 1-isothiocyanato-2-methyl-benzene, 2-methylphenylisothiocyanate, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, 2-methylbenzenisothiocyanate |
| PubChem CID | 69195 |
| IUPAC Name | 1-isothiocyanato-2-methylbenzene |
| SMILES | CC1=CC=CC=C1N=C=S |
Chemical Identifiers
| 614-69-7 | |
| 149.211 | |
| JYKYYPPZLPVIBY-UHFFFAOYSA-N | |
| 69195 | |
| CC1=CC=CC=C1N=C=S |
| C8H7NS | |
| MFCD00004802 | |
| 2-methylphenyl isothiocyanate, o-tolyl isothiocyanate, 2-tolyl isothiocyanate, o-tolylisothiocyanate, benzene, 1-isothiocyanato-2-methyl, 1-isothiocyanato-2-methyl-benzene, 2-methylphenylisothiocyanate, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, 2-methylbenzenisothiocyanate | |
| 1-isothiocyanato-2-methylbenzene |
Specifications
| 614-69-7 | |
| 239°C | |
| MFCD00004802 | |
| 2810 | |
| JYKYYPPZLPVIBY-UHFFFAOYSA-N | |
| 1-isothiocyanato-2-methylbenzene | |
| 69195 | |
| ≥99.0% (GC) | |
| o-Tolyl Isothiocyanate |
| Yellow | |
| C8H7NS | |
| 5 g | |
| 2-methylphenyl isothiocyanate, o-tolyl isothiocyanate, 2-tolyl isothiocyanate, o-tolylisothiocyanate, benzene, 1-isothiocyanato-2-methyl, 1-isothiocyanato-2-methyl-benzene, 2-methylphenylisothiocyanate, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, 2-methylbenzenisothiocyanate | |
| CC1=CC=CC=C1N=C=S | |
| 149.211 | |
| 149.21 | |
| Liquid |
Safety and Handling
TSCA : No