missing translation for 'onlineSavingsMsg'
Learn More

o-Tolyl isothiocyanate, 98%

Catalog No. p-7026694
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AAB2284506 5 g
AAB2284514 25 g
AAB2284522 100 g
3 options

Catalog No. AAB2284506

Supplier: Thermo Scientific Chemicals B2284506

Only null left

CAS: 614-69-7 | C8H7NS | 149.211 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 614-69-7
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00004802
InChI Key JYKYYPPZLPVIBY-UHFFFAOYSA-N
Synonym 2-methylphenyl isothiocyanate, o-tolyl isothiocyanate, 2-tolyl isothiocyanate, o-tolylisothiocyanate, benzene, 1-isothiocyanato-2-methyl, 1-isothiocyanato-2-methyl-benzene, 2-methylphenylisothiocyanate, isothiocyanic acid o-tolyl ester, isothiocyanic acid, o-tolyl ester, 2-methylbenzenisothiocyanate
PubChem CID 69195
IUPAC Name 1-isothiocyanato-2-methylbenzene
SMILES CC1=CC=CC=C1N=C=S

Specifications

Density 1.115
Boiling Point 239°C
Flash Point 107°C (224°F)
Odor Pungent
Refractive Index 1.6361
Quantity 5 g
UN Number UN2922
Beilstein 878487
Sensitivity Moisture sensitive
Formula Weight 149.22
Percent Purity 98%
Chemical Name or Material o-Tolyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H317-H334
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P342+P311-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 210-390-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only