missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Tolyl Isocyanate 98.0+%, TCI America™
Supplier: TCI America T032025G
Specifications
| o-Tolyl Isocyanate | |
| Yellow | |
| C8H7NO | |
| 25 g | |
| o-tolyl isocyanate, 2-methylphenyl isocyanate, 2-tolyl isocyanate, o-methylphenyl isocyanate, o-toluene isocyanate, benzene, 1-isocyanato-2-methyl, 2-methylphenylisocyanate, tolyl isocyanate, isocyanic acid, o-tolyl ester, o-tolylisocyanate | |
| CC1=CC=CC=C1N=C=O | |
| 133.15 | |
| CHEBI:60099 | |
| ≥98.0% (GC) |
| 614-68-6 | |
| 186°C | |
| MFCD00002009 | |
| 2206 | |
| VAYMIYBJLRRIFR-UHFFFAOYSA-N | |
| 1-isocyanato-2-methylbenzene | |
| 69194 | |
| 133.15 | |
| Liquid |
Chemical Identifiers
| 614-68-6 | |
| 133.15 | |
| VAYMIYBJLRRIFR-UHFFFAOYSA-N | |
| 69194 | |
| 1-isocyanato-2-methylbenzene |
| C8H7NO | |
| MFCD00002009 | |
| o-tolyl isocyanate, 2-methylphenyl isocyanate, 2-tolyl isocyanate, o-methylphenyl isocyanate, o-toluene isocyanate, benzene, 1-isocyanato-2-methyl, 2-methylphenylisocyanate, tolyl isocyanate, isocyanic acid, o-tolyl ester, o-tolylisocyanate | |
| CHEBI:60099 | |
| CC1=CC=CC=C1N=C=O |
Safety and Handling
EINECSNumber : (3)-3238
RTECSNumber : DA3678333
TSCA : Yes
Recommended Storage : Refrigerator