missing translation for 'onlineSavingsMsg'
Learn More

o-Tolyl Isocyanate 98.0+%, TCI America™

Catalog No. p-7136219
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
T03205G 5 g
T032025G 25 g
2 options

Catalog No. T03205G

Supplier: TCI America T03205G

Only null left

Chemical Identifiers

CAS 614-68-6
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00002009
InChI Key VAYMIYBJLRRIFR-UHFFFAOYSA-N
Synonym o-tolyl isocyanate, 2-methylphenyl isocyanate, 2-tolyl isocyanate, o-methylphenyl isocyanate, o-toluene isocyanate, benzene, 1-isocyanato-2-methyl, 2-methylphenylisocyanate, tolyl isocyanate, isocyanic acid, o-tolyl ester, o-tolylisocyanate
PubChem CID 69194
ChEBI CHEBI:60099
IUPAC Name 1-isocyanato-2-methylbenzene
SMILES CC1=CC=CC=C1N=C=O

Specifications

Color Yellow
Boiling Point 186°C
Quantity 5 g
UN Number 2206
Formula Weight 133.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material o-Tolyl Isocyanate
EINECSNumber (3)-3238
RTECSNumber DA3678333
TSCA Yes
Recommended Storage Refrigerator