Learn More
Chemical Identifiers
| CAS | 614-68-6 |
|---|---|
| Molecular Formula | C8H7NO |
| Molecular Weight (g/mol) | 133.15 |
| MDL Number | MFCD00002009 |
| InChI Key | VAYMIYBJLRRIFR-UHFFFAOYSA-N |
| Synonym | o-tolyl isocyanate, 2-methylphenyl isocyanate, 2-tolyl isocyanate, o-methylphenyl isocyanate, o-toluene isocyanate, benzene, 1-isocyanato-2-methyl, 2-methylphenylisocyanate, tolyl isocyanate, isocyanic acid, o-tolyl ester, o-tolylisocyanate |
| PubChem CID | 69194 |
| ChEBI | CHEBI:60099 |
| IUPAC Name | 1-isocyanato-2-methylbenzene |
| SMILES | CC1=CC=CC=C1N=C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC292220100
|
N/A | 10g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C8H7NO | |
| MFCD00002009 | |
| o-tolyl isocyanate, 2-methylphenyl isocyanate, 2-tolyl isocyanate, o-methylphenyl isocyanate, o-toluene isocyanate, benzene, 1-isocyanato-2-methyl, 2-methylphenylisocyanate, tolyl isocyanate, isocyanic acid, o-tolyl ester, o-tolylisocyanate | |
| CHEBI:60099 | |
| CC1=CC=CC=C1N=C=O |
Specifications
| 614-68-6 | |
| 185°C | |
| Authentic | |
| C8H7NO | |
| CH3C6H4NCO | |
| 12, 812 | |
| o-tolyl isocyanate, 2-methylphenyl isocyanate, 2-tolyl isocyanate, o-methylphenyl isocyanate, o-toluene isocyanate, benzene, 1-isocyanato-2-methyl, 2-methylphenylisocyanate, tolyl isocyanate, isocyanic acid, o-tolyl ester, o-tolylisocyanate | |
| CC1=CC=CC=C1N=C=O | |
| 133.15 | |
| CHEBI:60099 | |
| 99+% |
| 1.0740g/mL | |
| 92°C | |
| 99% min. (GC) | |
| 1.5335 to 1.5355 | |
| MFCD00002009 | |
| 1.074 | |
| VAYMIYBJLRRIFR-UHFFFAOYSA-N | |
| 1-isocyanato-2-methylbenzene | |
| 69194 | |
| 133.15 | |
| o-Tolyl isocyanate, 99+% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 210-389-0
RTECSNumber : DA3678333
TSCA : TSCA
Recommended Storage : Product may develop some turbidity or precipitate
RUO – Research Use Only