missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Tolyl isocyanate, 99+%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Density | 1.0740g/mL |
| Boiling Point | 185°C |
| Flash Point | 92°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5335 to 1.5355 |
| Linear Formula | CH3C6H4NCO |
| Quantity | 10g |
| Beilstein | 12, 812 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Fatal if inhaled. Causes severe skin burns and eye damage. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water |
| EINECSNumber | 210-389-0 |
| RTECSNumber | DA3678333 |
| TSCA | TSCA |
| Recommended Storage | Product may develop some turbidity or precipitate |
RUO – Research Use Only