missing translation for 'onlineSavingsMsg'
Learn More

o-Toluoyl Chloride 98.0+%, TCI America™

Catalog No. T031025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T031025G 25 g
T0310100G 100 g
T0310500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T031025G Supplier TCI America Supplier No. T031025G

Chemical Identifiers

CAS 933-88-0
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000668
InChI Key GPZXFICWCMCQPF-UHFFFAOYSA-N
Synonym o-toluoyl chloride, benzoyl chloride, 2-methyl, o-toluoylchloride, methylbenzoyl chloride, o-methylbenzoyl chloride, 2-toluoyl chloride, o-toluic acid chloride, unii-j9ykn18e7r, o-toluyl chloride, 2-methylbenzoic acid chloride
PubChem CID 70276
IUPAC Name 2-methylbenzoyl chloride
SMILES CC1=CC=CC=C1C(Cl)=O

Specifications

Color Yellow
Boiling Point 213°C
Quantity 25 g
UN Number 3265
Formula Weight 154.59
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material o-Toluoyl Chloride
EINECSNumber (3)-1399&(3)-2972
TSCA Yes