accessibility menu, dialog, popup

UserName

CH3ClO

CAS RN 933-88-0

IUPAC Name: 2-methylbenzoyl chloride
Synonyms: benzoyl chloride, 2-methyl-
Molecular Weight (g/mol): 154.59
Molecular Formula: C8H7ClO
InChi Key: GPZXFICWCMCQPF-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C(Cl)=O
Boiling Point 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (6)