missing translation for 'onlineSavingsMsg'
Learn More

o-Toluoyl chloride, 99%

Catalog No. AC153370050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153370050 5 g Glass bottle
AC153371000 100 g Glass bottle
AC153375000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC153370050 Supplier Thermo Scientific Chemicals Supplier No. 153370050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 933-88-0 | C8H7ClO | 154.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 933-88-0
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000668
InChI Key GPZXFICWCMCQPF-UHFFFAOYSA-N
Synonym o-toluoyl chloride, benzoyl chloride, 2-methyl, o-toluoylchloride, methylbenzoyl chloride, o-methylbenzoyl chloride, 2-toluoyl chloride, o-toluic acid chloride, unii-j9ykn18e7r, o-toluyl chloride, 2-methylbenzoic acid chloride
PubChem CID 70276
IUPAC Name 2-methylbenzoyl chloride
SMILES CC1=CC=CC=C1C(Cl)=O

Specifications

Color Pink to Yellow
Density 1.1800g/mL
Boiling Point 88°C to 90°C (12.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5540 to 1.556
Linear Formula CH3C6H4COCl
Quantity 5 g
Beilstein 09, 464
Specific Gravity 1.18
Solubility Information Solubility in water: decomposes
Formula Weight 154.6
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material o-Toluoyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-273-8
TSCA TSCA

RUO – Research Use Only