Learn More
o-Toluoyl chloride, 98+%, Thermo Scientific Chemicals
$116.51 - $116.51
Chemical Identifiers
| CAS | 933-88-0 |
|---|---|
| Molecular Formula | C8H7ClO |
| Molecular Weight (g/mol) | 154.59 |
| MDL Number | MFCD00000668 |
| InChI Key | GPZXFICWCMCQPF-UHFFFAOYSA-N |
| Synonym | o-toluoyl chloride, benzoyl chloride, 2-methyl, o-toluoylchloride, methylbenzoyl chloride, o-methylbenzoyl chloride, 2-toluoyl chloride, o-toluic acid chloride, unii-j9ykn18e7r, o-toluyl chloride, 2-methylbenzoic acid chloride |
| PubChem CID | 70276 |
| IUPAC Name | 2-methylbenzoyl chloride |
| SMILES | CC1=CC=CC=C1C(Cl)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1077522
|
Thermo Scientific Chemicals
A1077522 |
100 g |
Each for $116.51
|
|
|||||
Description
o-Toluoyl chloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationso-Toluoyl chloride is used as a pharmaceutical intermediate.
Solubility
Reacts with water.
Notes
Keep container tightly sealed. Store under dry inert gas. Store in cool, dry conditions in well sealed containers. Incompatible with bases, active metals, alcohols,amines and oxidizing agents. It is sensitive to moisture.
Chemical Identifiers
| 933-88-0 | |
| 154.59 | |
| GPZXFICWCMCQPF-UHFFFAOYSA-N | |
| 70276 | |
| CC1=CC=CC=C1C(Cl)=O |
| C8H7ClO | |
| MFCD00000668 | |
| o-toluoyl chloride, benzoyl chloride, 2-methyl, o-toluoylchloride, methylbenzoyl chloride, o-methylbenzoyl chloride, 2-toluoyl chloride, o-toluic acid chloride, unii-j9ykn18e7r, o-toluyl chloride, 2-methylbenzoic acid chloride | |
| 2-methylbenzoyl chloride |
Specifications
| 933-88-0 | |
| 212°C to 214°C | |
| Pungent | |
| 1.5549 | |
| 100 g | |
| 507933 | |
| o-toluoyl chloride, benzoyl chloride, 2-methyl, o-toluoylchloride, methylbenzoyl chloride, o-methylbenzoyl chloride, 2-toluoyl chloride, o-toluic acid chloride, unii-j9ykn18e7r, o-toluyl chloride, 2-methylbenzoic acid chloride | |
| GPZXFICWCMCQPF-UHFFFAOYSA-N | |
| 2-methylbenzoyl chloride | |
| 70276 | |
| ≥98% |
| 1.187 | |
| 76°C (168°F) | |
| C8H7ClO | |
| MFCD00000668 | |
| UN3265 | |
| Moisture sensitive | |
| Reacts with water. | |
| CC1=CC=CC=C1C(Cl)=O | |
| 154.59 | |
| 154.6 | |
| o-Toluoyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 213-273-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only