missing translation for 'onlineSavingsMsg'
Learn More

o-Toluidine 99.0+%, TCI America™

Catalog No. T029925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T029925G 25 g
T0299500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T029925G Supplier TCI America Supplier No. T029925G

Chemical Identifiers

CAS 95-53-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00007730
InChI Key RNVCVTLRINQCPJ-UHFFFAOYSA-N
Synonym o-toluidine, 2-toluidine, o-tolylamine, 2-aminotoluene, 2-methylbenzenamine, ortho-toluidine, o-methylaniline, benzenamine, 2-methyl, o-aminotoluene, o-methylbenzenamine
PubChem CID 7242
ChEBI CHEBI:66892
IUPAC Name 2-methylaniline
SMILES CC1=CC=CC=C1N

Specifications

Melting Point -28°C
Color Yellow
Boiling Point 201°C
Quantity 25 g
UN Number 1708
Formula Weight 107.16
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material o-Toluidine
EINECSNumber (3)-0186
RTECSNumber XU2975000
TSCA Yes