Learn More
o-Toluidine, 99%
CAS: 95-53-4 | C7H9N | 107.15 g/mol
Supplier: Thermo Scientific Chemicals 139080025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| o-Toluidine | |
| 95-53-4 | |
| Colorless to Yellow | |
| 199.0°C to 200.0°C | |
| Authentic | |
| Glass bottle | |
| 1.5710 to 1.5730 | |
| MFCD00007730 | |
| 12,772 | |
| 15,9696 | |
| Solubility in water: 1.5g/100mL (25°C). Other solubilities: solible in diluted acids,soluble in alcohol and ether,miscible with ethanol | |
| CC1=CC=CC=C1N | |
| 107.15 | |
| CHEBI:66892 | |
| 99% |
| 99% | |
| -23.0°C | |
| 1.0000g/mL | |
| 85°C | |
| 98.5% min. (GC) | |
| C7H9N | |
| CH3C6H4NH2 | |
| 2.5 L | |
| 1 | |
| o-toluidine, 2-toluidine, o-tolylamine, 2-aminotoluene, 2-methylbenzenamine, ortho-toluidine, o-methylaniline, benzenamine, 2-methyl, o-aminotoluene, o-methylbenzenamine | |
| RNVCVTLRINQCPJ-UHFFFAOYSA-N | |
| 2-methylaniline | |
| 7242 | |
| 107.15 | |
| Liquid |
Chemical Identifiers
| 95-53-4 | |
| 107.15 | |
| RNVCVTLRINQCPJ-UHFFFAOYSA-N | |
| 7242 | |
| 2-methylaniline |
| C7H9N | |
| MFCD00007730 | |
| o-toluidine, 2-toluidine, o-tolylamine, 2-aminotoluene, 2-methylbenzenamine, ortho-toluidine, o-methylaniline, benzenamine, 2-methyl, o-aminotoluene, o-methylbenzenamine | |
| CHEBI:66892 | |
| CC1=CC=CC=C1N |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye damage.
Toxic if inhaled.
Suspected of causing genetic defects if inhaled.
May cause cancer by inhalat
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cau
GHS Signal Word: Danger
EINECSNumber : 202-429-0
RTECSNumber : XU2975000
TSCA : TSCA
Recommended Storage : This substance darkens on storage
RUO – Research Use Only