missing translation for 'onlineSavingsMsg'
Learn More

o-Toluidine, 99%, Thermo Scientific Chemicals

Catalog No. AAA136020B
Change view
Click to view available options
Quantity:
1000 g
5000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA136020B 1000 g
AAA136020I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA136020B Supplier Thermo Scientific Chemicals Supplier No. A136020B
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

o-Toluidine is used in the production of dyes, as a presumptive test for blood in forensic science. And it is also used in determination of glucose in biological materials, as a precursor in the synthesis of poly(o-toluidine) and copper nanoparticle composite material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Toluidine is used in the production of dyes, as a presumptive test for blood in forensic science. And it is also used in determination of glucose in biological materials, as a precursor in the synthesis of poly(o-toluidine) and copper nanoparticle composite material.

Solubility
Soluble in water(15g/L).

Notes
Air & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 95-53-4
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00007730
InChI Key RNVCVTLRINQCPJ-UHFFFAOYSA-N
Synonym o-toluidine, 2-toluidine, o-tolylamine, 2-aminotoluene, 2-methylbenzenamine, ortho-toluidine, o-methylaniline, benzenamine, 2-methyl, o-aminotoluene, o-methylbenzenamine
PubChem CID 7242
ChEBI CHEBI:66892
IUPAC Name 2-methylaniline
SMILES CC1=CC=CC=C1N

Specifications

Melting Point -24°C
Density 1.004
Boiling Point 199°C to 200°C
Flash Point 85°C (185°F)
Odor Unpleasant
Refractive Index 1.572
Quantity 1000 g
UN Number UN1708
Beilstein 741981
Sensitivity Air and light sensitive
Merck Index 14,9536
Solubility Information Soluble in water(15g/L).
Formula Weight 107.16
Percent Purity 99%
Chemical Name or Material o-Toluidine
Show More Show Less
Hazard Category H227-H301+H331-H312-H315-H318-H341-H350
Hazard Statement GHS H Statement
H301-H331-H350-H227-H319
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.
Combustible liquid.
Causes serious eye irritation.
Precautionary Statement P201-P202-P210-P235-P261-P264b-P270-P271-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOLUIDINES, LIQUID
EINECSNumber 202-429-0
RTECSNumber XU2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only