missing translation for 'onlineSavingsMsg'
Learn More

o-Toluic Acid 98.0+%, TCI America™

Catalog No. t029225g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
T029225G 25 g
T0292100G 100 g
T0292500G 500 g
3 options

Catalog No. T029225G

Supplier: TCI America T029225G

Only null left

Chemical Identifiers

CAS 118-90-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002477
InChI Key ZWLPBLYKEWSWPD-UHFFFAOYSA-N
Synonym o-toluic acid, o-toluylic acid, orthotoluic acid, 2-toluic acid, toluic acid, benzoic acid, 2-methyl, o-methylbenzoic acid, 2-methyl-benzoic acid, benzoic acid, methyl, o-toluate
PubChem CID 8373
ChEBI CHEBI:36632
IUPAC Name 2-methylbenzoic acid
SMILES CC1=CC=CC=C1C(=O)O

Specifications

Melting Point 104°C
Color White-Yellow
Boiling Point 259°C
Quantity 25 g
Formula Weight 136.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material o-Toluic Acid
EINECSNumber (3)-1285
RTECSNumber XU1400000
TSCA Yes