missing translation for 'onlineSavingsMsg'
Learn More

o-Toluenesulfonyl Chloride (contains ca. 23% isomer) 75.0+%, TCI America™

Catalog No. T028425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T028425G 25 g
T0284500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T028425G Supplier TCI America Supplier No. T028425G

Chemical Identifiers

CAS 133-59-5
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.641
MDL Number MFCD00024874
InChI Key HDECRAPHCDXMIJ-UHFFFAOYSA-N
Synonym o-toluenesulfonyl chloride, 2-methylbenzene-1-sulfonyl chloride, toluenesulfonyl chloride, benzenesulfonyl chloride, 2-methyl, 2-toluenesulfonyl chloride, toluene-2-sulphonyl chloride, o-tolylsulfonyl chloride, o-tosyl chloride, unii-341l82e3wg, 2-methyl-benzenesulfonyl chloride
PubChem CID 8623
IUPAC Name 2-methylbenzenesulfonyl chloride
SMILES CC1=CC=CC=C1S(=O)(=O)Cl

Specifications

Color Yellow
Quantity 25 g
UN Number 3265
Formula Weight 190.64
Percent Purity ≥75.0% (GC)
Physical Form Liquid
Chemical Name or Material o-Toluenesulfonyl Chloride (contains ca. 23% isomer)
EINECSNumber (3)-1897
TSCA Yes