accessibility menu, dialog, popup

UserName

CH3SClOO

CAS RN 133-59-5

IUPAC Name: 2-methylbenzene-1-sulfonyl chloride
Synonyms: 2-methylbenzenesulfonyl chloride
Molecular Weight (g/mol): 190.64
Molecular Formula: C7H7ClO2S
InChi Key: HDECRAPHCDXMIJ-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1S(Cl)(=O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)