missing translation for 'onlineSavingsMsg'
Learn More

o-Toluenesulfonamide 98.0+%, TCI America™

Catalog No. T028025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T028025G 25 g
T0280500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T028025G Supplier TCI America Supplier No. T028025G

Chemical Identifiers

CAS 88-19-7
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.214
MDL Number MFCD00007934
InChI Key YCMLQMDWSXFTIF-UHFFFAOYSA-N
Synonym o-toluenesulfonamide, 2-methylbenzene-1-sulfonamide, benzenesulfonamide, 2-methyl, 2-toluenesulfonamide, o-methylbenzenesulfonamide, toluene-2-sulfonamide, ketjenflex 9, toluene-2-sulphonamide, ortho-toluenesulfonamide, 2-tolylsulfonamide
PubChem CID 6924
IUPAC Name 2-methylbenzenesulfonamide
SMILES CC1=CC=CC=C1S(=O)(=O)N

Specifications

Melting Point 155°C
Color White
Quantity 25 g
Formula Weight 171.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material o-Toluenesulfonamide
EINECSNumber (3)-1895
RTECSNumber XT4900000
TSCA Yes