Learn More
o-Toluenesulfonamide, 98%
CAS: 88-19-7 | C7H9NO2S | 171.214 g/mol
Supplier: Thermo Scientific Chemicals B2054418
Description
o-Toluene sulfonamide was mainly produced as a chemical intermediate for the production of saccharin. A mixture of o-Toluene sulfonamide with the p-isomer is used as a plasticizer for hot -melt adhesives, a chemical intermediate for fluorescent pigments and a chemical intermediate for plasticizer resins. (S080800). Studies suggest that o-Toluene sulfonamide has potential mutagenic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationso-Toluene sulfonamide was mainly produced as a chemical intermediate for the production of saccharin. A mixture of o-Toluene sulfonamide with the p-isomer is used as a plasticizer for hot -melt adhesives, a chemical intermediate for fluorescent pigments and a chemical intermediate for plasticizer resins. (S080800). Studies suggest that o-Toluene sulfonamide has potential mutagenic activity.
Solubility
Soluble in water (1.6 g/L at 25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases and acids.
Specifications
| o-Toluenesulfonamide | |
| 156°C | |
| 178°C (352°F) | |
| MFCD00007934 | |
| 1102362 | |
| Soluble in water (1.6g/L at 25°C). | |
| CC1=CC=CC=C1S(=O)(=O)N | |
| 171.214 | |
| 171.22 |
| 88-19-7 | |
| 210°C (1 mmHg) | |
| C7H9NO2S | |
| 50 g | |
| o-toluenesulfonamide, 2-methylbenzene-1-sulfonamide, benzenesulfonamide, 2-methyl, 2-toluenesulfonamide, o-methylbenzenesulfonamide, toluene-2-sulfonamide, ketjenflex 9, toluene-2-sulphonamide, ortho-toluenesulfonamide, 2-tolylsulfonamide | |
| YCMLQMDWSXFTIF-UHFFFAOYSA-N | |
| 2-methylbenzenesulfonamide | |
| 6924 | |
| 99% |
Chemical Identifiers
| 88-19-7 | |
| 171.214 | |
| YCMLQMDWSXFTIF-UHFFFAOYSA-N | |
| 6924 | |
| CC1=CC=CC=C1S(=O)(=O)N |
| C7H9NO2S | |
| MFCD00007934 | |
| o-toluenesulfonamide, 2-methylbenzene-1-sulfonamide, benzenesulfonamide, 2-methyl, 2-toluenesulfonamide, o-methylbenzenesulfonamide, toluene-2-sulfonamide, ketjenflex 9, toluene-2-sulphonamide, ortho-toluenesulfonamide, 2-tolylsulfonamide | |
| 2-methylbenzenesulfonamide |
Safety and Handling
GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P281-P308+P313-P501c
H351
EINECSNumber : 201-808-0
RTECSNumber : XT4900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only