missing translation for 'onlineSavingsMsg'
Learn More

o-Tolualdehyde 98.0+%, TCI America™

Catalog No. T0566500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0566500G 500 g
T056625G 25 g
T0566100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0566500G Supplier TCI America Supplier No. T0566500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 529-20-4
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00003338
InChI Key BTFQKIATRPGRBS-UHFFFAOYSA-N
Synonym o-tolualdehyde, 2-tolualdehyde, 2-formyltoluene, o-toluylaldehyde, o-toluic aldehyde, o-tolylaldehyde, o-methylbenzaldehyde, benzaldehyde, 2-methyl, 2-methyl-benzaldehyde, o-methylbenazldehyde
PubChem CID 10722
ChEBI CHEBI:27434
IUPAC Name 2-methylbenzaldehyde
SMILES CC1=CC=CC=C1C=O

Specifications

Color Yellow
Boiling Point 200°C
Quantity 500 g
Formula Weight 120.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material o-Tolualdehyde
EINECSNumber (3)-1143&(3)-2659
TSCA No