missing translation for 'onlineSavingsMsg'
Learn More

o-Tolualdehyde, 98%, stabilized

CAS: 529-20-4 | C8H8O | 120.15 g/mol

$122.78 - $122.78

Chemical Identifiers

CAS 529-20-4
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00003338
InChI Key BTFQKIATRPGRBS-UHFFFAOYSA-N
Synonym o-tolualdehyde, 2-tolualdehyde, 2-formyltoluene, o-toluylaldehyde, o-toluic aldehyde, o-tolylaldehyde, o-methylbenzaldehyde, benzaldehyde, 2-methyl, 2-methyl-benzaldehyde, o-methylbenazldehyde
PubChem CID 10722
ChEBI CHEBI:27434
IUPAC Name 2-methylbenzaldehyde
SMILES CC1=CC=CC=C1C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC152430250
SDS View Documents
Thermo Scientific Chemicals
152430250
25 g Glass bottle
Each for $122.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

529-20-4
120.15
BTFQKIATRPGRBS-UHFFFAOYSA-N
10722
2-methylbenzaldehyde
C8H8O
MFCD00003338
o-tolualdehyde, 2-tolualdehyde, 2-formyltoluene, o-toluylaldehyde, o-toluic aldehyde, o-tolylaldehyde, o-methylbenzaldehyde, benzaldehyde, 2-methyl, 2-methyl-benzaldehyde, o-methylbenazldehyde
CHEBI:27434
CC1=CC=CC=C1C=O

Specifications

529-20-4,123-31-9
Colorless to Yellow
199°C to 200°C
Authentic
Glass bottle
1.5460 to 1.548
MFCD00003338
07, 295
15, 9686
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol and ether
CC1=CC=CC=C1C=O
120.15
10722
120.15
Liquid
-35°C
1.0390g/mL
67°C
97.5% min. (GC)
C8H8O
CH3C6H4CHO
25 g
1.039
o-tolualdehyde, 2-tolualdehyde, 2-formyltoluene, o-toluylaldehyde, o-toluic aldehyde, o-tolylaldehyde, o-methylbenzaldehyde, benzaldehyde, 2-methyl, 2-methyl-benzaldehyde, o-methylbenazldehyde
BTFQKIATRPGRBS-UHFFFAOYSA-N
2-methylbenzaldehyde
0.1% hydroquinone
CHEBI:27434
98%
o-Tolualdehyde, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May produce an allergic reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen

GHS Signal Word: Warning

EINECSNumber : 208-452-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only