missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Tolidine Reagent, 0.135% (w/v) in 15% (v/v) Hydrochloric Acid, Ricca Chemical
Supplier: Ricca Chemical Company 86401
Specifications
| o-Tolidine Reagent | |
| 7732-18-5,7647-01-0,612-82-8 | |
| 87.0,15.32,0.12 | |
| 0.135% | |
| Amber Glass Bottle | |
| 15% (v/v) Hydrochloric Acid | |
| 4 L | |
| 0.135% (w/v) |
| Colorless to Green-Yellow | |
| 83.0,14.72,0.11 | |
| dihydrogen oxide, dihydrogen monoxide | |
| Laboratory | |
| APHA for Chlorine | |
| <1 | |
| Liquid |