missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Tolidine dihydrochloride, 97%
Description
o-Tolidine dihydrochloride is used as a sensitive colorimetric reagent for gold and for free chlorine in water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-Tolidine dihydrochloride is used as a sensitive colorimetric reagent for gold and for free chlorine in water.
Solubility
Soluble in water.
Notes
Air and light sensitive. Incompatible with oxidizing agents.
o-Tolidine dihydrochloride is used as a sensitive colorimetric reagent for gold and for free chlorine in water.
Solubility
Soluble in water.
Notes
Air and light sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Odor | Odorless |
| Quantity | 25 g |
| UN Number | UN3077 |
| Sensitivity | Air and light sensitive |
| Solubility Information | Soluble in water. |
| Formula Weight | 285.22 |
| Percent Purity | 97% |
| Chemical Name or Material | o-Tolidine dihydrochloride |
Safety and Handling
| Hazard Category | H302-H350 |
| Hazard Statement | GHS H Statement H350-H302 May cause cancer. Harmful if swallowed. |
| Precautionary Statement | P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 210-322-5 |
| RTECSNumber | DD1226000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only