missing translation for 'onlineSavingsMsg'
Learn More

O-tert-Butyl-N-Fmoc-L-beta-homotyrosine, 95%

Catalog No. AAH52192MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52192MD 250 mg
AAH5219203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52192MD Supplier Thermo Scientific Chemicals Supplier No. H52192MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 219967-69-8 | C29H31NO5 | 473.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 219967-69-8
Molecular Formula C29H31NO5
Molecular Weight (g/mol) 473.57
MDL Number MFCD01862862
InChI Key FKUBPXDMQFEPGP-CYLJNIGPNA-N
Synonym fmoc-l-beta-homotyrosine otbu, fmoc-o-t-butyl-l-beta-homotyrosine, fmoc-o-tert-butyl-l-beta-homotyrosine, fmoc-beta-homotyr tbu-oh, 3s-4-4-tert-butoxy phenyl-3-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, ambotzfaa6770, fmoc-homotyr tbu-oh, fmoc-beta-homotyr otbu-oh, tmba037, 3s-3-9h-fluoren-9-ylmethoxycarbonylamino-4-4-2-methylpropan-2-yl oxy phenyl butanoic acid
PubChem CID 2761556
SMILES CC(C)(C)OC1=CC=C(CC[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1

Specifications

Quantity 250 mg
Beilstein 8236890
IUPAC Name (2S)-4-[4-(tert-butoxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)butanoic acid
Formula Weight 473.57
Percent Purity 95%
Chemical Name or Material O-tert-Butyl-N-Fmoc-L-beta-homotyrosine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only