missing translation for 'onlineSavingsMsg'
Learn More

O-tert-Butyl-N-Fmoc-L-beta-homoserine, 95%

Catalog No. p-7034382
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52065MD 250 mg
AAH5206503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52065MD Supplier Thermo Scientific Chemicals Supplier No. H52065MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 203854-51-7 | C23H27NO5 | 397.471 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 203854-51-7
Molecular Formula C23H27NO5
Molecular Weight (g/mol) 397.471
MDL Number MFCD01862865
InChI Key SKYFRYZPXHNPOK-OAHLLOKOSA-N
Synonym fmoc-beta-hoser tbu-oh, fmoc-o-t-butyl-l-beta-homoserine, fmoc-l-beta-homoserine otbu, r-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy butanoic acid, 3r-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, ambotzfaa6740, fmoc-hoser tbu-oh, fmoc-??-hoser tbu-oh, fmoc-beta-homoser tbu-oh, fmoc-i 2-hoser tbu-oh
PubChem CID 2761544
IUPAC Name (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]butanoic acid
SMILES CC(C)(C)OCC(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 96°C to 98°C
Quantity 250 mg
Beilstein 8014825
Formula Weight 397.47
Percent Purity 95%
Chemical Name or Material O-tert-Butyl-N-Fmoc-L-beta-homoserine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only