missing translation for 'onlineSavingsMsg'
Learn More

O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine 98.0+%, TCI America™

Catalog No. p-7119997
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31685G 5 g
B316825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31685G Supplier TCI America Supplier No. B31685G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71989-33-8
Molecular Formula C22H24NO5
Molecular Weight (g/mol) 382.44
MDL Number MFCD00037127
InChI Key REITVGIIZHFVGU-IBGZPJMESA-M
Synonym fmoc-ser tbu-oh, fmoc-o-tert-butyl-l-serine, fmoc-ser t-bu-oh, fmoc-ser but, fmoc-o-tert.butyl-l-serine, fmoc-ser tbu, fmoc-ser but-oh, pubchem10035, fmoc-l-ser otbu oh, fmoc-l-ser tbu-oh
PubChem CID 2724633
IUPAC Name (2S)-3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoate
SMILES CC(C)(C)OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O

Specifications

Melting Point 136°C
Color White
Quantity 5 g
Formula Weight 383.44
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine