missing translation for 'onlineSavingsMsg'
Learn More

O-tert-Butyl-L-threonine tert-Butyl Ester 96.0+%, TCI America™

Catalog No. p-7122886
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33405G 5 g
B334025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33405G Supplier TCI America Supplier No. B33405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5854-78-4
Molecular Formula C12H25NO3
Molecular Weight (g/mol) 231.34
MDL Number MFCD00077109
InChI Key PPDIUNOGUIAFLV-UHFFFAOYNA-N
Synonym H-Thr(tBu)-OtBu
PubChem CID 7018832
IUPAC Name tert-butyl 2-amino-3-(tert-butoxy)butanoate
SMILES CC(OC(C)(C)C)C(N)C(=O)OC(C)(C)C

Specifications

Boiling Point 70°C
Quantity 5 g
Formula Weight 231.34
Percent Purity ≥96.0% (T)
Physical Form Liquid
Chemical Name or Material O-tert-Butyl-L-threonine tert-Butyl Ester