accessibility menu, dialog, popup

UserName

H3COOH3CCH3H3CNH2OCH3CH3H3C

CAS RN 5854-78-4

IUPAC Name: tert-butyl 2-amino-3-(tert-butoxy)butanoate
Synonyms: tert-butyl 2-amino-3-(tert-butoxy)butanoate
Molecular Weight (g/mol): 231.34
Molecular Formula: C12H25NO3
InChi Key: PPDIUNOGUIAFLV-UHFFFAOYNA-N
SMILES: CC(OC(C)(C)C)C(N)C(=O)OC(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results