missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-tert-Butyl-L-serine 97.0+%, TCI America™
$82.02 - $285.21
Chemical Identifiers
| CAS | 18822-58-7 |
|---|---|
| Molecular Formula | C7H15NO3 |
| Molecular Weight (g/mol) | 161.201 |
| MDL Number | MFCD00066088 |
| InChI Key | DDCPKNYKNWXULB-YFKPBYRVSA-N |
| Synonym | h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh |
| PubChem CID | 7004940 |
| IUPAC Name | (2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid |
| SMILES | CC(C)(C)OCC(C(=O)O)N |
Chemical Identifiers
| 18822-58-7 | |
| 161.201 | |
| DDCPKNYKNWXULB-YFKPBYRVSA-N | |
| 7004940 | |
| CC(C)(C)OCC(C(=O)O)N |
| C7H15NO3 | |
| MFCD00066088 | |
| h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh | |
| (2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid |
Specifications
| 18822-58-7 | |
| White | |
| MFCD00066088 | |
| h-ser tbu-oh, o-tert-butyl-l-serine, o-t-butyl-l-serine, s-2-amino-3-tert-butoxy propanoic acid, l-serine, o-1,1-dimethylethyl, 2s-2-amino-3-tert-butoxy propanoic acid, 2s-2-amino-3-2-methylpropan-2-yl oxy propanoic acid, ser tbu-oh, h-ser but-oh | |
| CC(C)(C)OCC(C(=O)O)N | |
| 161.201 | |
| 161.20 | |
| Crystalline Powder |
| 226°C | |
| C7H15NO3 | |
| 1 g | |
| DDCPKNYKNWXULB-YFKPBYRVSA-N | |
| (2S)-2-amino-3-[(2-methylpropan-2-yl)oxy]propanoic acid | |
| 7004940 | |
| ≥97.0% (HPLC,T) | |
| O-tert-Butyl-L-serine |
Safety and Handling
TSCA : No