missing translation for 'onlineSavingsMsg'
Learn More

O-TBDPS-D-Thr-N-Boc-L-tert-Leu-Diphenylphosphine 98.0+%, TCI America™

Catalog No. T2937100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2937100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2937100MG Supplier TCI America Supplier No. T2937100MG

Chemical Identifiers

CAS 1264520-63-9
Molecular Formula C43H57N2O4PSi
Molecular Weight (g/mol) 724.998
InChI Key XHYZJEQNKWDJPN-WLGXKCQKSA-N
Synonym N-(tert-Butoxycarbonyl)-L-tert-leucine N-[(2R,3S)-3-(tert-Butyldiphenylsilyloxy)-1-(diphenylphosphino)-2-butyl]amide, N-Boc-L-tert-leucine N-[(2R,3S)-3-(tert-Butyldiphenylsilyloxy)-1-(diphenylphosphino)-2-butyl]amide
PubChem CID 56954565
IUPAC Name tert-butyl N-[(2S)-1-[[(2R,3S)-3-[tert-butyl(diphenyl)silyl]oxy-1-diphenylphosphanylbutan-2-yl]amino]-3,3-dimethyl-1-oxobutan-2-yl]carbamate
SMILES CC(C(CP(C1=CC=CC=C1)C2=CC=CC=C2)NC(=O)C(C(C)(C)C)NC(=O)OC(C)(C)C)O[Si](C3=CC=CC=C3)(C4=CC=CC=C4)C(C)(C)C

Specifications

Melting Point 56°C
Color White
Quantity 100 mg
Formula Weight 725.00
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material O-TBDPS-D-Thr-N-Boc-L-tert-Leu-Diphenylphosphine