accessibility menu, dialog, popup

UserName

CH3PNHOCH3H3CCH3HNOOCH3CH3H3COSiCH3CH3H3C

CAS RN 1264520-63-9

IUPAC Name: tert-butyl N-[1-({3-[(tert-butyldiphenylsilyl)oxy]-1-(diphenylphosphanyl)butan-2-yl}carbamoyl)-2,2-dimethylpropyl]carbamate
Synonyms: tert-butyl N-[1-({3-[(tert-butyldiphenylsilyl)oxy]-1-(diphenylphosphanyl)butan-2-yl}carbamoyl)-2,2-dimethylpropyl]carbamate
Molecular Weight (g/mol): 725
Molecular Formula: C43H57N2O4PSi
InChi Key: XHYZJEQNKWDJPN-UHFFFAOYNA-N
SMILES: CC(O[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C(C)(C)C)C(CP(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)C(NC(=O)OC(C)(C)C)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results