missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Sulfobenzimide 99.0+%, TCI America™
$29.83 - $188.27
Chemical Identifiers
| CAS | 81-07-2 |
|---|---|
| Molecular Formula | C7H5NO3S |
| Molecular Weight (g/mol) | 183.181 |
| MDL Number | MFCD00005866 |
| InChI Key | CVHZOJJKTDOEJC-UHFFFAOYSA-N |
| Synonym | saccharin, saccharine, o-sulfobenzimide, o-benzoic sulfimide, saccharimide, benzosulfimide, o-benzosulfimide, benzoic sulfimide, benzosulphimide, saccharinose |
| PubChem CID | 5143 |
| ChEBI | CHEBI:32111 |
| IUPAC Name | 1,1-dioxo-1,2-benzothiazol-3-one |
| SMILES | C1=CC=C2C(=C1)C(=O)NS2(=O)=O |
Chemical Identifiers
| 81-07-2 | |
| 183.181 | |
| CVHZOJJKTDOEJC-UHFFFAOYSA-N | |
| 5143 | |
| 1,1-dioxo-1,2-benzothiazol-3-one |
| C7H5NO3S | |
| MFCD00005866 | |
| saccharin, saccharine, o-sulfobenzimide, o-benzoic sulfimide, saccharimide, benzosulfimide, o-benzosulfimide, benzoic sulfimide, benzosulphimide, saccharinose | |
| CHEBI:32111 | |
| C1=CC=C2C(=C1)C(=O)NS2(=O)=O |
Specifications
| 81-07-2 | |
| White | |
| MFCD00005866 | |
| saccharin, saccharine, o-sulfobenzimide, o-benzoic sulfimide, saccharimide, benzosulfimide, o-benzosulfimide, benzoic sulfimide, benzosulphimide, saccharinose | |
| C1=CC=C2C(=C1)C(=O)NS2(=O)=O | |
| 183.181 | |
| CHEBI:32111 | |
| ≥99.0% (HPLC,T) | |
| o-Sulfobenzimide |
| 226°C | |
| C7H5NO3S | |
| 25 g | |
| CVHZOJJKTDOEJC-UHFFFAOYSA-N | |
| 1,1-dioxo-1,2-benzothiazol-3-one | |
| 5143 | |
| 183.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0198
RTECSNumber : DE4200000
TSCA : Yes