missing translation for 'onlineSavingsMsg'
Learn More

o-Sulfobenzimide 99.0+%, TCI America™

Catalog No. B0004500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0004500G 500 g
B000425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0004500G Supplier TCI America Supplier No. B0004500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 81-07-2
Molecular Formula C7H5NO3S
Molecular Weight (g/mol) 183.181
MDL Number MFCD00005866
InChI Key CVHZOJJKTDOEJC-UHFFFAOYSA-N
Synonym saccharin, saccharine, o-sulfobenzimide, o-benzoic sulfimide, saccharimide, benzosulfimide, o-benzosulfimide, benzoic sulfimide, benzosulphimide, saccharinose
PubChem CID 5143
ChEBI CHEBI:32111
IUPAC Name 1,1-dioxo-1,2-benzothiazol-3-one
SMILES C1=CC=C2C(=C1)C(=O)NS2(=O)=O

Specifications

Melting Point 226°C
Color White
Quantity 500 g
Formula Weight 183.18
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material o-Sulfobenzimide
EINECSNumber (5)-0198
RTECSNumber DE4200000
TSCA Yes