missing translation for 'onlineSavingsMsg'
Learn More

o-Sulfobenzimide Sodium Salt Dihydrate 98.0+%, TCI America™

Catalog No. B013125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B013125G 25 g
B0131500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B013125G Supplier TCI America Supplier No. B013125G

Chemical Identifiers

CAS 6155-57-3
Molecular Formula C7H9NNaO5S
Molecular Weight (g/mol) 242.201
MDL Number MFCD00151213
InChI Key UQKRDMFNUSMUCB-UHFFFAOYSA-N
Synonym saccharin sodium dihydrate, saccharin sodium, sodium saccharin, saccharin soluble, saccharin sodium salt hydrate, 1,2-benzisothiazol-3 2h-one, 1,1-dioxide, sodium salt, dihydrate, o-sulfobenzimide sodium salt, 2-sulfobenzoic acid imide sodium salt, saccharin sodium salt dihydrate, c7h4nnao3s.2h2o
PubChem CID 46942257
IUPAC Name 1,1-dioxo-1,2-benzothiazol-3-one;sodium;dihydrate
SMILES C1=CC=C2C(=C1)C(=O)NS2(=O)=O.O.O.[Na]

Specifications

Melting Point 228°C
Color White
Quantity 25 g
Formula Weight 205.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material o-Sulfobenzimide Sodium Salt Dihydrate
EINECSNumber (5)-0199
RTECSNumber DE4550000
TSCA Yes