missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Succinyl-L-carnitine lithium salt ≥98.0% (TLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| Linear Formula | C11H19 NO6 · xLi+ |
| Type | Organic Standard |
| Quantity | 10 mg |
| Formula Weight | 261.27 (free acid basis) |
| Percent Purity | ≥98% (TLC) |