missing translation for 'onlineSavingsMsg'
Learn More

o-Phthalaldehyde 99.0+%, TCI America™

Supplier:  TCI America P02805G

 View more versions of this product

Catalog No. P02805G


Only null left

Description

Description

Specifications

Specifications

o-Phthalaldehyde [for HPLC Labeling]
84°C
C8H6O2
2811
ZWLUXSQADUDCSB-UHFFFAOYSA-N
benzene-1,2-dicarbaldehyde
4807
134.13
5 g
56°C
643-79-8
MFCD00003335
Phthaldialdehyde, Benzene-1,2-dicarboxaldehyde
O=CC1=CC=CC=C1C=O
134.13
CHEBI:70851
≥99.0% (GC)
Crystal/Powder

Chemical Identifiers

643-79-8
134.13
ZWLUXSQADUDCSB-UHFFFAOYSA-N
4807
benzene-1,2-dicarbaldehyde
C8H6O2
MFCD00003335
Phthaldialdehyde, Benzene-1,2-dicarboxaldehyde
CHEBI:70851
O=CC1=CC=CC=C1C=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1145

RTECSNumber : TH6950000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions