missing translation for 'onlineSavingsMsg'
Learn More

o-Phosphoric Acid, 85% (Certified ACS), Fisher Chemical™

Catalog No. p-20451
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Flint Glass Bottle
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A242500 500 mL Flint Glass Bottle
A242212 2.5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A242500 Supplier Thermo Fisher Scientific Supplier No. BPA242 500
Only null left

Chemical Identifiers

CAS 7664-38-2 , 7732-18-5
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

pH 1
Melting Point 21°C
Boiling Point 158°C
Density 1.680g/cm³
Heavy Metals (as Pb) 0.001% max.
Identification Pass Test
Packaging Flint Glass Bottle
Reducing Substances Pass Test
Vapor Pressure 2 hPa at 20°C
Normality about 44.6
Assay Percent Range ≥85.0 w/w %
Quantity 500 mL
Physical Form Liquid
UN Number 1805
Formula Weight 98
Percent Purity ≥85.0% (w/w)
Grade Certified ACS
Chemical Name or Material o-Phosphoric Acid, 85%
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.

NFPA

Health:3
Flammability:0
Instability:0
DOTInformation DOT Class 8, : Corrosive