missing translation for 'onlineSavingsMsg'
Learn More

O-Phospho-L-tyrosine 97.0+%, TCI America™

Catalog No. P0959100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0959100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0959100MG Supplier TCI America Supplier No. P0959100MG
Only null left

Chemical Identifiers

CAS 21820-51-9
Molecular Formula C9H12NO6P
Molecular Weight (g/mol) 261.17
MDL Number MFCD00002603
InChI Key DCWXELXMIBXGTH-QMMMGPOBSA-N
PubChem CID 30819
ChEBI CHEBI:37788
IUPAC Name (2S)-2-amino-3-[4-(phosphonooxy)phenyl]propanoic acid
SMILES N[C@@H](CC1=CC=C(OP(O)(O)=O)C=C1)C(O)=O

Specifications

Color White
Quantity 100 mg
Formula Weight 261.17
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material O-Phospho-L-tyrosine
TSCA No
Recommended Storage Freezer (-20 °C)