accessibility menu, dialog, popup

UserName

CAS RN 21820-51-9

H2N(S)OPHOOHOHOO

CAS RN 21820-51-9

IUPAC Name: (2S)-2-amino-3-[4-(phosphonooxy)phenyl]propanoic acid
Synonyms: phosphonotyrosine
Molecular Weight (g/mol): 261.17
Molecular Formula: C9H12NO6P
InChi Key: DCWXELXMIBXGTH-QMMMGPOBSA-N
SMILES: N[C@@H](CC1=CC=C(OP(O)(O)=O)C=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results