missing translation for 'onlineSavingsMsg'
Learn More

O-Phenylhydroxylamine hydrochloride, 97%

Catalog No. AC438810010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC438810010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC438810010 Supplier Thermo Scientific Chemicals Supplier No. 438810010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6092-80-4 | C6H7NO·ClH | 145.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6092-80-4
Molecular Formula C6H7NO·ClH
Molecular Weight (g/mol) 145.59
InChI Key DBTXKJJSFWZJNS-UHFFFAOYSA-N
Synonym o-phenylhydroxylamine hydrochloride, o-phenylhydroxylamine hcl, phenoxyamine hydrochloride, hydroxylamine, o-phenyl-, hydrochloride, acmc-20ak08, n-phenoxyamine hydrochloride, o-phenylhydroxylamne hydrochlorde, o-phenyl-hydroxylamine hydrochloride, o-phenylhydroxylamine hydrochloride at
PubChem CID 6365179
IUPAC Name O-phenylhydroxylamine;hydrochloride
SMILES C1=CC=C(C=C1)ON.Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 121.0°C to 122.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 145.59
Percent Purity 97%
Physical Form Powder
Chemical Name or Material O-Phenylhydroxylamine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
If skin irritation occurs: Get medical advice/attention.
If eye irritatio
EINECSNumber 228-039-

RUO – Research Use Only