missing translation for 'onlineSavingsMsg'
Learn More

o-Phenylenediamine, 98%

CAS: 95-54-5 | C6H8N2 | 108.14 g/mol

Supplier:  Thermo Scientific Chemicals 130550010

 View more versions of this product

Catalog No. AC130550010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

o-Phenylenediamine
97.5
100.0°C to 103.0°C
256.0°C to 258.0°C
Authentic
Glass bottle
C6H4(NH2)2
1 kg
01,834; 08,393
o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine
GEYOCULIXLDCMW-UHFFFAOYSA-N
benzene-1,2-diamine
7243
108.14
Crystalline Powder
95-54-5
100.0
White to Beige
136°C
97.5% min. (GC)
C6H8N2
MFCD00007721
13, IV, 38
15, 7396
Solubility in water: 54g/L (20°C). Other solubilities: soluble in alcohol,ether and chloroform
C1=CC=C(C(=C1)N)N
108.14
CHEBI:34043
98%

Chemical Identifiers

95-54-5
108.14
GEYOCULIXLDCMW-UHFFFAOYSA-N
7243
benzene-1,2-diamine
C6H8N2
MFCD00007721
o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine
CHEBI:34043
C1=CC=C(C(=C1)N)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Suspected of causing cancer.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause an allergic skin reaction.
Suspected of causing genetic

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a posit

GHS Signal Word: Danger

EINECSNumber : 202-430-6

Recommended Storage : This product tends to darken during storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only