missing translation for 'onlineSavingsMsg'
Learn More

o-Phenylenediamine, 98%

Catalog No. AAA1194630
Change view
Click to view available options
Quantity:
25 g
250 g
1000 g
5000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1194630 250 g
AAA1194614 25 g
AAA119460B 1000 g
AAA119460I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1194630 Supplier Thermo Scientific Chemicals Supplier No. A1194630
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-54-5 | C6H8N2 | 108.144 g/mol

o-phenylenediamine is used to prepare tiabendazole, pyrazinamide, morinamide, clemizole and chlormidazole. It is involved in the synthesis of benzimidazole by reacting with carboxylic acids and their derivatives. It is also used to prepare quinoxalinedione and mercaptoimidazoles by condensation with dimethyl oxalate and xanthate esters respectively. It reacts with nitrous acid to get benzotriazole, which finds application as a corrosion inhibitor. It is actively involved in the preparation of pharmaceuticals by reacting substituted o-phenylenediamine with various diketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-phenylenediamine is used to prepare tiabendazole, pyrazinamide, morinamide, clemizole and chlormidazole. It is involved in the synthesis of benzimidazole by reacting with carboxylic acids and their derivatives. It is also used to prepare quinoxalinedione and mercaptoimidazoles by condensation with dimethyl oxalate and xanthate esters respectively. It reacts with nitrous acid to get benzotriazole, which finds application as a corrosion inhibitor. It is actively involved in the preparation of pharmaceuticals by reacting substituted o-phenylenediamine with various diketones.

Solubility
Soluble in alcohol, chloroform, ether, slightly soluble in water. Soluble in hot water.

Notes
Moisture and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 95-54-5
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00007721
InChI Key GEYOCULIXLDCMW-UHFFFAOYSA-N
Synonym o-phenylenediamine, 1,2-phenylenediamine, 1,2-benzenediamine, 1,2-diaminobenzene, 2-aminoaniline, o-diaminobenzene, orthamine, o-benzenediamine, opda, ortho-phenylenediamine
PubChem CID 7243
ChEBI CHEBI:34043
IUPAC Name benzene-1,2-diamine
SMILES C1=CC=C(C(=C1)N)N

Specifications

Melting Point 100°C to 103°C
Density 1.270
Boiling Point 256°C to 258°C
Flash Point 136°C (277°F)
Assay Percent Range 98%
Quantity 250 g
UN Number UN1673
Beilstein 606074
Sensitivity Air Sensitive
Merck Index 14,7284
Solubility Information Soluble in alcohol,chloroform,ether,slightly soluble in water; Soluble in hot water.
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material o-Phenylenediamine
Show More Show Less
Hazard Category H301-H312+H332-H317-H319-H341-H351-H500
Hazard Statement GHS H Statement
H301-H341-H351-H312-H332-H319-H317
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: PHENYLENEDIAMINES
EINECSNumber 202-430-6
RTECSNumber SS7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only