missing translation for 'onlineSavingsMsg'
Learn More

o-Phenetidine 98.0+%, TCI America™

Catalog No. p-7134419
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P008925G 25 g
P0089500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P008925G Supplier TCI America Supplier No. P008925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 94-70-2
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00007689
InChI Key ULHFFAFDSSHFDA-UHFFFAOYSA-N
Synonym o-phenetidine, 2-aminophenetole, benzenamine, 2-ethoxy, o-ethoxyaniline, 2-ethoxybenzenamine, o-aminophenetole, 2-ethoxyphenylamine, ethoxyaniline, 2-phenetidine, benzenamine, ar-ethoxy
PubChem CID 7203
IUPAC Name 2-ethoxyaniline
SMILES CCOC1=CC=CC=C1N

Specifications

Melting Point -4°C
Color Yellow
Boiling Point 232°C
Quantity 25 g
UN Number 2311
Formula Weight 137.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material o-Phenetidine
EINECSNumber (3)-0682&(3)-2773
RTECSNumber SI6465300
TSCA Yes