missing translation for 'onlineSavingsMsg'
Learn More

o-Phenetidine, 99%

Catalog No. AC233480050
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC233480050 5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC233480050 Supplier Thermo Scientific Chemicals Supplier No. 233480050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 94-70-2 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94-70-2
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00007689
InChI Key ULHFFAFDSSHFDA-UHFFFAOYSA-N
Synonym o-phenetidine, 2-aminophenetole, benzenamine, 2-ethoxy, o-ethoxyaniline, 2-ethoxybenzenamine, o-aminophenetole, 2-ethoxyphenylamine, ethoxyaniline, 2-phenetidine, benzenamine, ar-ethoxy
PubChem CID 7203
IUPAC Name 2-ethoxyaniline
SMILES CCOC1=CC=CC=C1N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -20.0°C
Color Red
Density 1.0500g/mL
Boiling Point 230.0°C to 233.0°C
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5540 to 1.5560
Linear Formula C2H5OC6H4NH2
Quantity 5 mL
Specific Gravity 1.05
Merck Index 15, 7341
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 137.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material o-Phenetidine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-356-4
RTECSNumber SI6465300
TSCA TSCA

RUO – Research Use Only